Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Solvent Delivery Caps with Inlet Valve and Filter

Catalog No. 13311010
Change view
Click to view available options
Size:
GL38
GL45
Nalgene 38-430
Wako 38 mm
Material:
Black/white PTFE, PP, SS, EPDM
Black/white PTFE, PP, SS, FFKM
Black/white PTFE, PP, SS, Viton
Blue/white PTFE, PP, SS, Viton
Green/white PTFE, PP, SS, EPDM
White PTFE, PP, SS, EPDM
White PTFE, PP, SS, FFKM
White PTFE, PP, SS, Viton
Yellow/white PTFE, PP, SS, FFKM
Type:
Four 1/4 in.-28 ports
Two 1/4 in.-28 ports
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Type
13-311-010 GL45 Blue/white PTFE, PP, SS, Viton Two 1/4 in.-28 ports
13-311-006 GL45 Green/white PTFE, PP, SS, EPDM Two 1/4 in.-28 ports
13-311-007 GL45 Green/white PTFE, PP, SS, EPDM Four 1/4 in.-28 ports
13-311-008 GL45 Yellow/white PTFE, PP, SS, FFKM Two 1/4 in.-28 ports
13-311-009 GL45 Yellow/white PTFE, PP, SS, FFKM Four 1/4 in.-28 ports
13-311-011 GL45 Blue/white PTFE, PP, SS, Viton Four 1/4 in.-28 ports
13-311-018 GL38 Black/white PTFE, PP, SS, FFKM Two 1/4 in.-28 ports
13-311-019 GL38 Black/white PTFE, PP, SS, EPDM Two 1/4 in.-28 ports
13-311-020 GL38 Black/white PTFE, PP, SS, Viton Two 1/4 in.-28 ports
13-311-021 GL38 Black/white PTFE, PP, SS, FFKM Four 1/4 in.-28 ports
13-311-022 GL38 Black/white PTFE, PP, SS, EPDM Four 1/4 in.-28 ports
13-311-023 GL38 Black/white PTFE, PP, SS, Viton Four 1/4 in.-28 ports
13-311-030 Nalgene 38-430 White PTFE, PP, SS, EPDM Two 1/4 in.-28 ports
13-311-031 Nalgene 38-430 White PTFE, PP, SS, EPDM Four 1/4 in.-28 ports
13-311-032 Nalgene 38-430 White PTFE, PP, SS, FFKM Two 1/4 in.-28 ports
13-311-033 Nalgene 38-430 White PTFE, PP, SS, FFKM Four 1/4 in.-28 ports
13-311-034 Nalgene 38-430 White PTFE, PP, SS, Viton Two 1/4 in.-28 ports
13-311-035 Nalgene 38-430 White PTFE, PP, SS, Viton Four 1/4 in.-28 ports
13-311-042 Wako 38 mm White PTFE, PP, SS, EPDM Two 1/4 in.-28 ports
13-311-044 Wako 38 mm White PTFE, PP, SS, FFKM Two 1/4 in.-28 ports
13-311-045 Wako 38 mm White PTFE, PP, SS, FFKM Four 1/4 in.-28 ports
13-311-046 Wako 38 mm White PTFE, PP, SS, Viton Two 1/4 in.-28 ports
13-311-047 Wako 38 mm White PTFE, PP, SS, Viton Four 1/4 in.-28 ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 13-311-010 Supplier Cole-Parmer™ Supplier No. 1201810
Add to Cart
Add to Cart

VapLock™ Solvent Delivery Caps with Inlet Valve and Filter reduce chemical evaporation and exposure to hazardous vapors

Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) OD

Warranty and Services

None

Specifications

For Use With (Equipment) Solvent-Compatible Containers
Material Blue/white PTFE, PP, SS, Viton
Size GL45
Thread Size (English) 1/4 in.-28 UNF(F)
For Use With (Application) Solvent Safety
Type Two 1/4 in.-28 ports
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.