Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
Description
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | Solvent-Compatible Containers |
| Material | Polypropylene |
| Size | Nalgene 38-430 |
| Thread Size (English) | 1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F) |
| For Use With (Application) | Solvent Safety |
| Type | Solvent Waste Kit |