Description
- Various cap sizes available to fit your solvent container
- 10 port manifold—1 x 0.5 in. NPT(F), 3 x 0.25 in. NPT(F), and 6 x 1/4-28 UNF(F)
- Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers.
- Waste kits include a 10-port manifold, exhaust filter, adapters, plugs, and tubing—everything you need to install on a typical HPLC instrument.
- Manifold allows solvent waste to be collected from multiple effluent streams.
- Exhaust filter minimize exposure to harmful chemical vapors.
- Fittings and tubing provide leak-tight connections minimizing spills and leaks.
Specifications
Specifications
| Material | Polypropylene |
| Size | 63 mm Fort-Pak Container |
| Thread Size | 1 x 0.5 in. NPT(F), 3 x 0.25 in. NPT(F), and 6 x 1/4-28 UNF(F) |
| Product Type | Solvent Waste Kit |