Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Drums and Panel Mounting
Catalog No. 13311168
Change view
Click to view available options
Size:
2 in. Coarse Thread
2 in. Fine Thread
Panel Mount
Material:
PTFE
Polypropylene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material
13-311-168 2 in. Fine Thread PTFE
13-311-167 2 in. Fine Thread Polypropylene
13-311-173 2 in. Coarse Thread Polypropylene
13-311-179 Panel Mount Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-311-168 Supplier Cole-Parmer™ Supplier No. 1201966
Add to Cart
Add to Cart
VapLock™ Solvent Waste Kits for Drums and Panel Mounting collect effluent from multiple sources while minimizing harmful solvent exhaust
Various cap sizes available to fit your solvent container
Warranty and Services
None
Specifications
For Use With (Equipment) Solvent-Compatible Containers
Material PTFE
Size 2 in. Fine Thread
Thread Size (English) 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
For Use With (Application) Solvent Safety
Type Solvent Waste Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov