Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails
VapLock™ Solvent Waste Kits for Jars and Pails collect effluent from multiple sources while minimizing harmful solvent exhaust
Supplier:  Cole-Parmer™ 1201923
 View more versions of this product
Catalog No. 13-311-125
Only null left
Add to Cart
Description
Description
Various cap sizes available to fit your solvent container
Warranty and Services
None
Specifications
Specifications
Solvent Safety
Solvent-Compatible Containers
70 mm Rieke
PTFE
Solvent Waste Kit
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.