Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Justrite™ Containers
Description
Warranty and Services
None
Specifications
Specifications
| Closure Size | 1/2 in. NPT(F), 1/4 in. NPT(F) |
| Closure Type | Justrite Quick-Disconnect |
| For Use With (Application) | Solvent Safety |
| For Use With (Equipment) | Solvent-Compatible Containers |
| Material | Polypropylene, Stainless Steel Fitting |
| Product Type | Solvent Waste Manifold |