Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Panel Mounting
Description
- Fits panel aperture of 1-1/4 in and require a 1-3/4 in diameter clearance around the aperture
- Different materials, numbers and sizes of ports help customize to your application
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | Solvent-Compatible Containers |
| Material | Polypropylene |
| Size | Panel Mount |
| Thread Size (English) | 1/2 in. NPT(F), 1/4 in. NPT(F) |
| For Use With (Application) | Solvent Safety |
| Type | Solvent Waste Manifold |