Learn More
Cole-Parmer™ VapLock™ Threaded Plugs
Catalog No. 13311250
Change view
Click to view available options
Material:
POM
PTFE
Polypropylene
Thread Size (English):
1/2 in. NPT(M)
1/4 in. NPT(M)
1/4 in.-28 UNF(M)
1/8 in. NPT(M)
3/8 in. NPT(M)
5/16 in.-24 UNF(M)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Thread Size (English)
13-311-250 PTFE 1/4 in. NPT(M)
13-311-249 PTFE 1/4 in.-28 UNF(M)
13-311-253 PTFE 5/16 in.-24 UNF(M)
13-311-246 Polypropylene 1/4 in. NPT(M)
13-311-252 Polypropylene 1/8 in. NPT(M)
13-311-247 Polypropylene 1/2 in. NPT(M)
13-311-251 Polypropylene 3/8 in. NPT(M)
13-311-248 POM 1/4 in.-28 UNF(M)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 13-311-250 Supplier Cole-Parmer™ Supplier No. 1202048
Add to Cart
Add to Cart
Use VapLock™ Threaded Plugs to seal unused ports for a leak-resistant system
Seal unused manifold and bottle cap ports
Warranty and Services
None
Specifications
For Use With (Equipment) VapLock Solvent Containment Systems
Material PTFE
Thread Size (English) 1/4 in. NPT(M)
For Use With (Application) Solvent Safety
Type Port Plug