Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Threaded Plugs

Catalog No. 13311251
Change view
Click to view available options
Material:
POM
PTFE
Polypropylene
Thread Size (English):
1/2 in. NPT(M)
1/4 in. NPT(M)
1/4 in.-28 UNF(M)
1/8 in. NPT(M)
3/8 in. NPT(M)
5/16 in.-24 UNF(M)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Thread Size (English)
13-311-251 Polypropylene 3/8 in. NPT(M)
13-311-249 PTFE 1/4 in.-28 UNF(M)
13-311-253 PTFE 5/16 in.-24 UNF(M)
13-311-246 Polypropylene 1/4 in. NPT(M)
13-311-252 Polypropylene 1/8 in. NPT(M)
13-311-250 PTFE 1/4 in. NPT(M)
13-311-247 Polypropylene 1/2 in. NPT(M)
13-311-248 POM 1/4 in.-28 UNF(M)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 13-311-251 Supplier Cole-Parmer™ Supplier No. 1202049
Add to Cart
Add to Cart

Use VapLock™ Threaded Plugs to seal unused ports for a leak-resistant system

Seal unused manifold and bottle cap ports

Warranty and Services

None

Specifications

For Use With (Equipment) VapLock Solvent Containment Systems
Material Polypropylene
Thread Size (English) 3/8 in. NPT(M)
For Use With (Application) Solvent Safety
Type Port Plug
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.