Description
- Reduces fumes and harmful vapor exhaust
- Easily connect to Justrite™ Centura safety containers
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | Container Ports with 1/2 in. NPT(F) |
| Material | HDPE, Polypropylene |
| Thread Size | 1/2 in. NPT(M) |
| For Use With (Application) | Solvent Safety |
| Product Type | Waste Funnel, 4 in. dia |