Learn More
SOMNI Scientific™ Vaporizer Turnaround (Service and Return), Indicate: Type, Agent, Fill, Mount and Serial Number
SDP
Catalog No. 04777163 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-163 Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-163 Supplier SOMNI Scientific™ Supplier No. VTURNAROUND
Add to Cart
Add to Cart
Maintains device accuracy, proper function and prevents exposure from leaks. Send your vaporizer in for full service and calibration; receive your vaporizer back. SOMNI™ offers service on a wide range of anesthetic vaporizers.
  • Maintains accuracy
  • Prevents exposure from leaks
  • Full Service and Calibration
    • Certificate of Calibration included
  • Agents include Isoflurane and Sevoflurane
  • Fill types include Funnel and Key Fill
  • Tec 3, Tec 4, Tec 5
  • Draeger 19.1, 19.3, 2000
  • Ohio
  • Penlon
  • Extended capacity 3EX
  • 6 psi 
  • Service interval based on manufacturer recommendation
Specifications
For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) RM-17003, RM-17004, AMD-17004, TT-17000, OC-21003, OC-21004
Material Brass and Alloys
Model Agents include Isoflurane and Sevoflurane
Includes Calibration Certificate
For Veterinary Use Only