Learn More
Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit, Four Ports 1/8 in., Three Ports 1/4 in., and One Port 1/4 in. Or 3/8 in. Hose Barb
Shop All Justrite Safety Group Products

Description
- For packaging group II and hazardous chemicals
- Carbon filter prevents volatile organic compound vapors
- Two-piece cap design
Specifications
Specifications
| Capacity (English) | 3.6 gal. |
| Capacity (Metric) | 13.5 L |
| Certifications/Compliance | UN/DOT |
| Closure Material | Polypropylene |
| Closure Size | 70 mm |
| Color | White |
| Description | Four (4) 1/8 in. Tubing OD Ports, Three (3) 1/4 in. Tubing OD Ports and One (1) 1/4 or 3/8 in. Tubing ID Hose Barb Ports |
| Dimensions (L x W x H) | 287 x 235 x 333 mm |
| For Use With (Application) | Safe transport of hazardous liquids |
| Height (English) | 13.1 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.