Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit, Four Ports 1/8 in. and Three Ports 1/4 in.
GREENER_CHOICE

Catalog No. 19830396 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (Metric):
13.5 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19-830-396 13.5 L
19-830-397 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-830-396 Supplier Justrite™ Supplier No. 12842
Add to Cart
Add to Cart

For Use With: Safe transport of hazardous liquids; Closure Material: Polypropylene; Color: White

  • For packaging group II and hazardous chemicals
  • Carbon filter prevents volatile organic compound vapors
  • Two-piece cap design

Specifications

Capacity (English) 3.6 gal.
Capacity (Metric) 13.5 L
Certifications/Compliance UN/DOT
Closure Material Polypropylene
Closure Size 70 mm
Color White
Description Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Dimensions (L x W x H) 287 x 235 x 333 mm
For Use With (Application) Safe transport of hazardous liquids
Height (English) 13.1 in.
Length (English) 11.3 in.
Material HDPE
Product Type UN/DOT Carboy with Filter Kit
Shape Rectangular
Width (English) 9.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.