Learn More
Bachem Vapreotide Acetate
SDP
Supplier:  Bachem 4062833.0025
Encompass_Preferred
Vapreotide Acetate, H-D-Phe-Cys-Tyr-D-Trp-Lys-Val-Cys-Trp-NH2, 103222-11-3, C57H70N12O9S2, Mr 1131.39, 25mg. RC-160. Vapreotide is a potent octapeptide analog of somatostatin with selectivity for the somatostatin receptor subtypes SSTR-2 and SSTR-5. It is under investigation in the management of various disorders, including esophageal variceal bleeding, gastrointestinal and pancreatic fistulas, acromegaly, carcinoid tumors, and for the prevention of postoperative complications following pancreatic surgery.
Catalog No. 50-194-6728
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.