Learn More
Grainger Variable Frequency Drive 3 hp 460V AC
Supplier:  Grainger 14H067
Encompass_Preferred
The Grainger Approved Variable Frequency Drive is a compact AC drive for variable speed control of 3-phase AC induction motors. It features an easy-to-use keypad and display, a voltage range of 208-230V AC / 60 Hz, and current ratings of 2.3A to 17A. It includes DC link reactors to reduce harmonics and has programmable parameters. Designed for variable torque applications like fans and pumps, it offers energy savings by matching motor speed to load. The compact design allows easy installation in tight spaces.
Catalog No. 50-275-0656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.