Learn More
Environmental Express™ Variable Speed Metering Pump
Catalog No. 0973524 Shop All Environmental Express Products
Change view
Click to view available options
Material:
Ceramic, Ryton
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
09-735-24 Ceramic, Ryton
09-735-20 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-735-24 Supplier Environmental Express™ Supplier No. TP1200
Fluid metering pump with PTFE and Stainless Steel Construction
  • To connect to the Environmental Express Zero Headspace Extractor, Millipore or Gelman ZHE, you will need to purchase the pump adapter kit as well
  • Complies with EPA 1311
Specifications
Material Ceramic, Ryton
Voltage 115 V