Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Variable Volume, Fully Autoclavable Micropipettes
SDPSCI_ED

Catalog No. S43749 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2.0 μL
0.5 to 10 μL
0.5 to 5 mL
10 to 100 μL
100 to 1000 μL
2 to 10 mL
2 to 20 μL
20 to 200 μL
5 to 50 μL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S43749 5 to 50 μL
S43746 0.2 to 2.0 μL
S43747 0.5 to 10 μL
S43748 2 to 20 μL
S43750 10 to 100 μL
S43751 20 to 200 μL
S43752 100 to 1000 μL
S43753 0.5 to 5 mL
S43754 2 to 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. S43749 Supplier United Scientific™ Supplier No. PRV50
Add to Cart
Add to Cart

These Variable Volume, Fully Autoclavable Micropipettes are highly accurate and easy to calibrate/maintain.

  • Built-in tip ejectors and autoclavable tip cones work on a click stop digital system.
  • Easy to disassemble prior to autoclaving.
  • Manufactured in an ISO 9001:2008 certified facility.
  • Pipettes are individually calibrated according to ISO 8655 standards.
  • A calibration certificate is provided with each pipette.

Specifications

Autoclavable Autoclavable
Calibrated Yes
Description Calibrated According to ISO 8655 Standards
Material Plastic
Sterility Non-sterile
For Use With (Application) For use with 200 μL Tips
Graduated No
Graduations 1 μL
Graduation Type Digital
Includes Calibration Certificate
Color Blue
Volume (Metric) 5 to 50 μL
Length (Metric) 228 mm
Type Volumetric Pipet
Disposable Reusable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.