Learn More
United Scientific™ Variable Volume, Fully Autoclavable Micropipettes

Shop All United Scientific Supplies, Inc Products


Description
- Built-in tip ejectors and autoclavable tip cones work on a click stop digital system.
- Easy to disassemble prior to autoclaving.
- Manufactured in an ISO 9001:2008 certified facility.
- Pipettes are individually calibrated according to ISO 8655 standards.
- A calibration certificate is provided with each pipette.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Calibrated | Yes |
| Description | Calibrated According to ISO 8655 Standards |
| Material | Plastic |
| Sterility | Non-sterile |
| For Use With (Application) | For use with 5 mL tips |
| Graduated | No |
| Graduations | 100 μL |
| Graduation Type | Digital |
| Includes | Calibration Certificate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.