Learn More
Invitrogen™ Varicella zoster Virus ORF47 Polyclonal Antibody

Description
Varicella zoster Virus ORF47 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Varicella zoster Virus ORF47 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene Alias | varicella zoster virus ge |
| Host Species | Rabbit |
| Immunogen | Synthesized peptide derived from human KR2_VZVD. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.