Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam VAS 2870, NADPH Oxidase Inhibitor, Solid, 10 mg

Catalog No. 502705395
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5395 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5395 Supplier Abcam Supplier No. AB14520910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent NADPH oxidase inhibitor that blocks PDGF-mediated activation and ROS production.

  • Molecular weight: 360.4 g/mol
  • Purity: >98%
  • Form: Solid
  • Activity: NADPH oxidase inhibitor; blocks PDGF-mediated NADPH activation and ROS production
  • Potency: Inhibits PMS-induced oxidative burst (IC50 = 2 μM, HeLa cells)
  • Applications: Shows anti-ischemic effects ex vivo
  • Composition: 100% 2-(3-benzyltriazolo[4,5-d]pyrimidin-7-yl)sulfanyl-1,3-benzoxazole

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 2-(3-benzyltriazolo[4,5-d]pyrimidin-7-yl)sulfanyl-1,3-benzoxazole
Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Target Molecule NADPH Oxidase
Formulation 100% 2-(3-benzyltriazolo[4,5-d]pyrimidin-7-yl)sulfanyl-1,3-benzoxazole
Quantity 10 mg
For Use With (Application) NADPH oxidase inhibition, PDGF-mediated NADPH activation blocking, ROS production inhibition, oxidative burst inhibition, anti-ischemic research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.