Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam VAS 2870, NADPH Oxidase Inhibitor, Solid, 50 mg

Catalog No. 502705394
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5394 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5394 Supplier Abcam Supplier No. AB14520950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent NADPH oxidase inhibitor that blocks PDGF-mediated activation and ROS production.

  • Function: NADPH oxidase inhibitor
  • Molecular Weight: 360.4 g/mol
  • Purity: >98%
  • Form: Solid
  • Activity: Blocks PDGF-mediated NADPH activation and ROS production. Inhibits PMS-induced oxidative burst (IC50 = 2 μM, HeLa cells). Shows anti-ischemic effects ex vivo.
  • Storage: Store at ambient temperature.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule NADPH Oxidase
Content And Storage Store at ambient temperature.
Form Solid
Quantity 50 mg
For Use With (Application) NADPH oxidase inhibition, PDGF-mediated NADPH activation, ROS production, oxidative burst inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.