Learn More
Aviva Systems Biology Corp VASH1 Antibody - N-terminal region (ARP55099_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55099_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VASH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VASH1. Alternate names: KIAA1036ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61423