Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Vasoactive Intestinal Peptide (10-28), Human, Porcine, Rat, Ovine

Catalog No. AAJ66099EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66099EXD 0.5 mg
AAJ66099MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66099EXD Supplier Thermo Scientific™ Supplier No. J66099EXD

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Species Human, Porcine, Rat, Ovine
Purification Method HPLC
Common Name VIP (10-28), human, porcine, rat, ovine
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2338.84
Product Type Vasoactive Intestinal Peptide (10-28), human, porcine, rat, ovine
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sequence Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.