Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ VasoTACS In Situ Apoptosis Detection Kit
SDP

Catalog No. 482630K
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
482630K 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 482630K Supplier R&D Systems™ Supplier No. 482630K

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Cytonin™, 5mL; Proteinase K, 30uL; TACS Blue Label, 3mL; TACS-Nuclease, 15uL; Red Counterstain C, 50mL; Strep-HRP, 30uL; Also includes TdT enzyme, buffers and dNTPs, 50X Mn+2

Provides all the reagents for the detection of DNA fragmentation among vascular cells and tissue sections using TUNEL

Apoptosis is characterized by a number of intracellular phenomena such as membrane blebbing, chromatin condensation and nuclear DNA fragmentation. Detection of nuclear DNA fragmentation is a widely accepted method to assay for apoptosis and can be perform

Contain TACS BlueLabel, Red Counterstain C, and reagents necessary for generating positive controls
Kits are optimized for detection of apoptosis in vascular tissues
Study fixed cells, embedded tissues and frozen tissues using light microscopy
VasoTACS

Specifications

Quantity 30 Tests
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.