Learn More
Novus Biologicals™ VasoTACS™ In Situ Apoptosis Kit [TACS Blue Label]
Shop All Novus Biologicals Products
Description
- Generates intense blue staining in cells with DNA fragmentation
- Easily visualized against pale red counterstain
- Low background
Detection: Biotinylated dNTP/Streptavidin-HRP/Blue Label
Testing Format: Light Microscopy
Sample Type: Fixed cells, Embedded tissues, Frozen tissues
Specifications
Specifications
| Quantity | 30 Tests |
| Product Type | Apoptosis detection kit |