Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VAV2 Monoclonal Antibody (3F5D3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA538657
Description
This antibody has been tested in indirect ELISA.
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
VAV2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P52735 | |
Vav2 | |
Purified recombinant fragment of human VAV2 (aa: 552-868) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
3F5D3 | |
Unconjugated | |
Vav2 | |
2810040F13Rik; AI847175; Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; vav guanine nucleotide exchange factor 2; VAV2; VAV-2; Vav2 oncogene | |
Mouse | |
Protein G | |
RUO | |
7410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction