Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VBP1 Monoclonal Antibody (OTI2A3)
GREENER_CHOICE

Catalog No. PIMA526008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526008 Supplier Invitrogen™ Supplier No. MA526008
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VBP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by VBP1 interacts with the Von Hippel-Lindau protein to form an intracellular complex. The encoded protein functions as a chaperone protein, and may play a role in the transport of the Von Hippel-Lindau protein from the perinuclear granules to the nucleus or cytoplasm. Alternative splicing and the use of alternate transcription start sites results in multiple transcript variants encoding different protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Vbp1
Gene Accession No. P61758, P61759
Gene Alias EGK_21124; HIBBJ46; LOC703257; PFD3; Pfdn3; phosphatidylinositol-binding clathrin assembly protein; prefoldin 3; prefoldin subunit 3; prefoldin subunit 3; phosphatidylinositol-binding clathrin assembly protein; RP13-228J13.4; VBP1; VBP-1; VHL binding protein 1; VHL binding protein-1; VHL-binding protein 1; von Hippel-Lindau binding protein 1; von Hippel-Lindau-binding protein 1
Gene Symbols Vbp1
Host Species Mouse
Immunogen Full length human recombinant protein of VBP1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22327, 481089, 681825, 7411
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.