Description
- Compatible with VACSTAR digital and VACSTAR lite vacuum pumps
- Manual and programmable modes
- Valve cleaning function for long service life
- High energy efficiency thanks to device-to-device communication: control of the circulator by connecting the vacuum controller with PC 1.3 data cable (available as accessory)
- Integrated ventilation valve for simple pressure equalization and aeration with inert gas after process completion
- VENT short key button allows pressure equalization during a running process
- Program function: up to ten user-defined programs can be saved, with up to ten pressure/time sequences
- USB / RS232 interfaces for connection with the laboratory software labworldsoft™
- Electronic control system for improved process efficiency and higher solvent recovery rate
- High energy efficiency thanks to device-to-device communication (vacuum controller to recirculating chiller)
Specifications
Specifications
| Material | PBT |
| Type | Vacuum Controller/Interface |