Learn More
LABSYSTEMS DIAGNOSTICS OY VC-CALIBRATOR LYOPHILIZED 5 X 0,5 ML
Supplier:  LABSYSTEMS DIAGNOSTICS OY 72500405
Encompass_Preferred
VC-calibration standard is intended to be used in clinical laboratories for the quantitative determination of vitamin C in serum and plasma to detect deficiencies or hypervitaminosis. The calibrators are prepared using human serum and in a lyophilized form
Catalog No. NC2865592