Learn More
LABSYSTEMS DIAGNOSTICS OY VC-CONTROL SET 2 X 5 X 0,5ML
Supplier:  LABSYSTEMS DIAGNOSTICS OY 72500402
Encompass
VC-Control set (Low + High Control) is intended to be used in clinical laboratories for the quantitative determination of vitamin C in serum and plasma to detect deficiencies or hypervitaminosis. The controls are prepared using human serum and in a lyophilized form
Catalog No. NC2865591