Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VCAM-1 (CD106) Recombinant Mouse Monoclonal Antibody (B-K9 (C106-K9))
GREENER_CHOICE

Mouse Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA548283

Catalog No. PIMA548283


Only null left
Add to Cart

Description

Description

Specificity: B-K9 antibody recognizes CD106 (VCAM1). CD106 is a member of the Ig superfamily. It is a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis.

VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VCAM-1 (CD106)
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P19320
VCAM1
This antibody was raised by immunizing Balb/c mice against activated human umbilical vein endothelial cells (HUVEC).
200 μg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Inhibition Assays
B-K9 (C106-K9)
Unconjugated
VCAM1
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B
Mouse
Protein A
RUO
7412
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.