Learn More
Invitrogen™ VCAM-1 Monoclonal Antibody (1.4C3)
Description
Use on formalin-fixed sections requires boiling sections in 1 mM EDTA, pH 8.0 for 10-20 minutes followed by cooling at room temperature for 20 minutes. A suggested positive control for this product is tonsil tissue.
Specifications
Specifications
| Antigen | VCAM-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1.4C3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | VCAM1 |
| Gene Accession No. | P19320 |
| Gene Alias | adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.