Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse VCAM-1 ELISA Kit

Catalog No. EMVCAM1X10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMVCAM1X10 10 x 96 Tests
EMVCAM1 96 Tests
EMVCAM1X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EMVCAM1X10 Supplier Invitrogen™ Supplier No. EMVCAM1X10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Mouse VCAM-1 ELISA quantitates Ms VCAM-1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms VCAM-1. Principle of the method The Mouse VCAM-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29533
Assay Range 2.05 to 500 pg/mL
Assay Sensitivity 1.5 pg/mL
Calibration Range 1.5 pg/mL to 500 pg/mL
Description VCAM-1 ELISA Kit, 96 Assays, 1 Kit, Mouse Species, Streptavidin-HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of VCAM-1 in. Samples Using a Microplate Reader
Detectable Analytes VCAM-1
Assay Solid-phase sandwich ELISA
Conjugate HRP
Detection Method Colorimetric
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse 6 Ckine, CTACK, Eotaxin, GCSF, GM-CSF, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p40, IL-12p70, IL-13, IL-17, IFN-gamma, KC, Leptin, MCP-5, MIP-1 alpha, MIP-2, MIP-3 bet
Sufficient For 96 Assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 22329
Gene Symbol CD106,Vcam-1
Host Species Mouse
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym CD106, Vcam-1
Target VCAM-1 (CD106)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.