Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCIP135 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7141832
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3848 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3848 Supplier Bethyl Laboratories, Inc Supplier No. A302934AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Acts as a deubiquitinating enzyme. Necessary for VCP-mediated reassembly of Golgi stacks after mitosis. May play a role in VCP-mediated formation of transitional endoplasmic reticulum (tER). Mediates dissociation of the ternary complex containing STX5A, NSFL1C and VCP. Hydrolyzes ′Lys-11′- and ′Lys-48′-linked polyubiquitin chains.

Specifications

Antigen VCIP135
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene VCPIP1
Gene Accession No. Q8CDG3, Q96JH7
Gene Alias DUBA3, VCIP135, deubiquitinating protein VCIP135, valosin-containing protein (p97)/p47 complex-interacting protein p135, valosin-containing protein p97/p47 complex-interacting protein 1, valosin-containing protein p97/p47 complex-interacting protein p135
Gene Symbols VCPIP1
Host Species Rabbit
Immunogen Between 1172 and 1222
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70675, 80124
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.