Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam VCP Peptide, Synthetic, Mouse, >90% Purity, 50 μg

Catalog No. 502710981
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0981 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0981 Supplier Abcam Supplier No. AB3978850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >90% purity for neutralization and blocking applications.

  • Protein Type: Synthetic
  • Target: VCP (Valosin-containing protein)
  • Species Reactivity: Mouse
  • Purity: >90%
  • Applications: Neutralization (Neut), Blocking (BL)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule VCP (Mouse)
Substrate Properties Peptide-based Substrate
Product Type Blocker and Diluent
Concentration 0.5 mg/mL
Content And Storage Store at -20°C.
Form Liquid
Label or Dye Unconjugated
Format Vial
Cross Reactivity Mouse
Quantity 50 μg
For Use With (Application) Blocking, Neutralization
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.