Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Sodium Fluoride/Potassium Oxalate Additive, 4 mL, Hemogard™ Closure Plastic Tube, 100/Box
Supplier:  Med Vet International 367922
Encompass_Preferred
Features Sodium Fluoride, 10.0 mg / Potassium Oxalate, 8.0 mg For glucose determination
Catalog No. 50-304-3050
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.