Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VDAC2/VDAC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675765
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-765 Supplier Invitrogen™ Supplier No. PA141205
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human brain cell lysate, antigen standard for VDAC2 (transient overexpression lysate), human brain protein. 92% sequence identity with Mouse, Rat, Chicken and Zebrafish proteins. Immunogen sequence: KKSGKIKSSY KRE.

The Voltage-Dependant Anion Channel (VDAC) of the outer mitochondrial membrane, a small abundant protein found in all eukaryotic kingdoms, forms a voltage-gated pore when incorporated into planar lipid bilayers. VDAC is also the site of binding of the metabolic enzymes hexokinase and glycerol kinase to the mitochondrion in what may be a significant metabolic regulatory interaction. VDAC3 is involved specifically in translocation of adenine nucleotides through the outer membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VDAC2/VDAC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene VDAC2
Gene Accession No. P45880, P68002, Q9MZ13, Q9Y277
Gene Alias FLJ23841; HD-VDAC3; hVDAC-2; hVDAC-3; outer mitochondrial membrane protein porin 2; Outer mitochondrial membrane protein porin 3; POR; RP11-375G3.1; VDAC1P5; VDAC2; VDAC-2; VDAC2 protein; VDAC3; VDAC-3; VDAC5P; voltage dependent anion channel 2; voltage dependent anion channel 3; voltage-dependent anion channel 1 pseudogene 5; voltage-dependent anion channel 2; voltage-dependent anion channel 3; voltage-dependent anion channel 5, pseudogene; voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 3
Gene Symbols VDAC2, VDAC3
Host Species Rabbit
Immunogen Amino acids 120-132 of human VDAC2 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101092793, 101099117, 282120, 282716, 479255, 606963, 7417, 7419
Target Species Human, Canine, Monkey, Bovine, Feline
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.