Learn More
Enzo Life Sciences VE-cadherin polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6610100
Encompass_Preferred
Immunogen: Recombinant human VE-cadherin (aa 68-307). Species reactivity: Human. Applications: ICC, IF, IHC, IP, WB. Purity: Purified by antigen-specific affinity chromatography followed by protein A affinity chromatography. Formulation: Liquid. In 0.01M PBS, pH 7.4, containing 0.02% sodium azide and 50% glycerol.
Catalog No. 50-201-3185
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.