Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ VectaFluor(TM) Duet DL594 DL488 Labeling Kit
SDP

Catalog No. NB402713 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402713 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402713 Supplier Novus Biologicals™ Supplier No. DK8828NB
Add to Cart
Add to Cart

offers maximum convenience to achieve immunofluorescence double labeling in one step.

Specifications

For Use With (Application) Immunocytochemistry/ Immunofluorescence
Quantity 1 Kit
Content And Storage Store at 4°C. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.