Learn More
R&D Systems™ VectaFluor(TM) Excel Amp. anti-Mouse IgG DL488 Ab Kit
Shop All Novus Biologicals Products

Specifications
Specifications
| For Use With (Application) | Immunocytochemistry/ Immunofluorescence |
| Quantity | 1 Kit |
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.