Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ VectaFluor(TM) Excel Amp. anti-Mouse IgG DL594 Ab Kit
SDP

Catalog No. NB402715 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402715 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402715 Supplier R&D Systems™ Supplier No. DK2594NB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. offers a non-biotin amplification method for fluorescence applications.

Specifications

For Use With (Application) Immunocytochemistry/ Immunofluorescence
Quantity 1 Kit
Content And Storage Store at 4°C. Do not freeze.
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.