Learn More
R&D Systems™ VectaMount(R) AQ Aqueous Mounting Medium
SDP
Catalog No. NB398954 Shop All Novus Products
Change view
Click to view available options
Quantity:
60 mL
Catalog No. Quantity
NB398954 60 mL
1 options
Catalog No. NB398954 Supplier R&D Systems™ Supplier No. H550160NB
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. hardsetting mounting medium developed for use with enzymatic substrates, such as AEC, ImmPACT™ AEC, and ImmPACT AMEC Red, whose reaction products are soluble in alcohol or other organic solvents.
Specifications
Quantity 60 mL
Content And Storage Store at room temperature.
Product Line Novus