Learn More
Cell Biolabs Inc pAAV-MCS Promoterless Expression Vector, 10 µg
Supplier:  Cell Biolabs Inc VPK411
Encompass_Preferred
Cell Biolabs' pAAV-MCS Promoterless Expression Vector allows you to clone your gene along with a suitable promoter into the multiple cloning sites, and then co-transfect with AAV packaging plasmids into 293 cells to generate recombinant AAV. The cloning capacity is 3.9 kb.
Catalog No. 50-673-822