Specifications
| Certifications/Compliance | ANSI Z89.1-1997 Type II G and E; CSA 294.1-1992 Class E |
| Color | Forest Green |
| Material | Shell: HDPE, Core: Polystyrene Foam |
| Suspension Type | 4-Point Ratchet |