Learn More
Bullard™ Vector™ Helmet
SDP
Catalog No. 19034392 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Blue
Forest Green
Hi-vis Orange
Red
White
Yellow
Suspension Type:
4-Point Ratchet
4-Point Standard
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-034-392 Yellow 4-Point Ratchet
19-088-778C Blue 4-Point Ratchet
19-034-390 Hi-vis Orange 4-Point Ratchet
19-088-778A Red 4-Point Ratchet
19-034-391 Forest Green 4-Point Ratchet
19-088-778B White 4-Point Ratchet
19-088-777B White 4-Point Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-034-392 Supplier Bullard™ Supplier No. VTYLR
Specifications
Certifications/Compliance ANSI Z89.1-1997 Type II G and E; CSA 294.1-1992 Class E
Color Yellow
Material Shell: HDPE, Core: Polystyrene Foam
Suspension Type 4-Point Ratchet