Learn More
Cell Biolabs Inc pBABEpuro Retroviral Cloning Vector, 10 µg
Supplier:  Cell Biolabs Inc RTV001PURO
Encompass_Preferred
Clone your gene of interest into Cell Biolabs' pBABE-puro Retroviral Vector and then package into a retroviral particle using a retroviral packaging cell or co-transfect with retroviral packaging constructs into 293T cells.
Catalog No. NC0130601
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.