Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ VEGF 164 Antibody Pair [HRP]
SDP

Catalog No. NB010277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010277 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010277 Supplier Novus Biologicals™ Supplier No. NBP27961915PLATES
Only null left
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Rat VEGF 164.

Specifications

Assay Range 23.44 pg/mL
Assay Sensitivity 23.44-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Rat VEGF 164.
Target Species Rat
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 7422
Gene Symbol VEGFA
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-VEGF 164 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Rat VEGF 164 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 32 ng of recombinant Rat VEGF 164.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins
    Storage Requirements Storage is content dependent.
    Target VEGF 164
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.