Learn More
Description
Specifications
Specifications
| Assay Range | 15.6 - 1,000pg/mL |
| Assay Sensitivity | 7.9pg/mL |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | The assay detects both natural and recombinant human VEGF-A. Cross reactivity and interference of circulating factors of the immune system were evaluated by spiking these proteins at physiologically relevant concentrations into a human VEGF-A positive serum. There was no crossreactivity detected, notably not with human VEGF-B, VEGF-C, VEGF-D and PfGF. Interference was detected for VEGF-R1 at concentrations > 200 pg/ml, and not for VEGF-R2. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7422 |
| Gene Symbol | VEGFA |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human VEGF-A, Biotin-Conjugate anti-human VEGF-A polyclonal antibody, Streptavidin-HRP, Human VEGF-A Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
