Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Monoclonal Antibody (VG1), Invitrogen™

Catalog No. PIMA512184 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512184 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512184 Supplier Thermo Scientific Supplier No. MA512184
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA5-12184 targets Vascular Endothelial Growth Factor in IF, IHC (P) and WB applications and shows reactivity with Human samples.

VEGF (vascular endothelial growth factor) which is a 45 kDa homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. VEGF has a variety of effects on vascular endothelium, including the ability to promote endothelial cell viability, mitogenesis, chemotaxis, and vascular permeability. The VEGF family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis, and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells. VEGF and its high-affinity binding receptors, the tyrosine kinases FLK1 and FLT1, are thought to be important for the development of embryonic vasculature. Studies have shown that an alternately spliced form of FLT1 produces a soluble protein, termed sFLT1, which binds vascular endothelial growth factor with high affinity, playing an inhibitory role in angiogenesis. Elevated levels of VEGF is linked to POEMS syndrome (Polyneuropathy, Organomegaly, Endocrinopathy, Monoclonal gammopathy, Skin changes) also known as Crow-Fukase syndrome which affects multiple organs in the body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VG1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% sodium azide, pH 7.4
Gene VEGFA
Gene Accession No. P15692
Gene Alias angiogenic factor; etID309976.16; eVEGF120; eVEGF164; human VEGF121 homolog; MGC70609; MVCD1; ovine vascular endothelial growth factor; RP1-261G23.1; unnamed protein product; VAS; vascular endothelial growth factor; vascular endothelial growth factor (VEGF) precursor; vascular endothelial growth factor 121 isoform; vascular endothelial growth factor 164; vascular endothelial growth factor 165b; vascular endothelial growth factor 182; vascular endothelial growth factor 182 isoform; vascular endothelial growth factor 188; vascular endothelial growth factor 188 isoform; vascular endothelial growth factor 189; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular endothelial growth factor Aa; Vascular endothelial growth factor A-A; vascular endothelial growth factor isoform 165; vascular permeability factor; Vasculotropin; VEGF; VEGF1; VEGF164; VEGF182; VEGF188; vegfa; VEGF-A; VEGFA165b; vegfaa; VEGF-A-A; VEGFMGC70609; VPF; VPF ant; wu:fj82c06
Gene Symbols VEGFA
Host Species Mouse
Immunogen Recombinant VEGF189 protein
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.