Learn More
Genscript Corporation VEGF R2/KDRBiotin HisAvi
Supplier:  Genscript Corporation Z03970500
Encompass_Preferred
Tyrosine-protein kinase that acts as a cell-surface receptor for VEGFA VEGFB and PGF and plays an essential role in the development of embryonic vasculature the regulation of angiogenesis cell survival cell migration macrophage function chemotaxis and cancer cell invasion The tyrosine kinase receptor vascular endothelial growth factor receptor 2 (VEGFR2) is a key regulator of angiogenesis
Catalog No. 50-000-58340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.